Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GME | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203488.1 | NP_001190417.1 | 6 - 7 | 1172 | 91 | |
NM_122767.4 | NP_198236.1 | 6 - 7 | 1196 | 91 | ||
GenBank mRNA | AK226220.1 | - | 6 - 7 | 1152 | 91 | |
AY057660.2 | - | 6 - 7 | 1037 | 91 | ||
AY057694.1 | - | 6 - 7 | 1037 | 91 | ||
AY116953.1 | - | 5 - 6 | 974 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |