Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122950.4 | NP_198409.1 | 3 - 4 | 371 | 93 | |
GenBank mRNA | AK228466.1 | - | 3 - 4 | 303 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |