Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122951.1 | NP_198410.1 | 2 - 3 | 956 | 76 | |
GenBank mRNA | AF510166.1 | - | 2 - 3 | 956 | 76 | |
DQ447001.1 | - | 2 - 3 | 956 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |