Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122951.1 | NP_198410.1 | 4 - 5 | 1187 | 70 | |
GenBank mRNA | AF510166.1 | - | 4 - 5 | 1187 | 70 | |
DQ447001.1 | - | 4 - 5 | 1187 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |