Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122961.4 | NP_198420.1 | 13 - 14 | 1942 | 72 | |
GenBank mRNA | AY045780.1 | - | 13 - 14 | 1920 | 72 | |
AY142570.1 | - | 12 - 13 | 1717 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |