Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122968.2 | NP_198426.1 | 1 - 2 | 292 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |