Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBD35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122981.4 | NP_198439.1 | 1 | 470 | 122 | |
GenBank mRNA | AB473860.1 | - | 1 | 470 | 122 | |
DQ056696.1 | - | 1 | 453 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |