Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344283.1 | NP_001332027.1 | 13 - 14 | 1447 | 100 | |
NM_123266.8 | NP_198720.2 | 13 - 14 | 1511 | 100 | ||
GenBank mRNA | AK229810.1 | - | 13 - 14 | 1393 | 100 | |
DQ469798.1 | - | 13 - 14 | 1311 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |