Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344355.1 | NP_001332644.1 | 10 - 11 | 3253 | 98 | |
NM_001344356.1 | NP_001332643.1 | 10 - 11 | 3250 | 98 | ||
NM_001344357.1 | NP_001332642.1 | 10 - 11 | 3250 | 98 | ||
NM_123447.3 | NP_198898.2 | 10 - 11 | 3250 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |