Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344355.1 | NP_001332644.1 | 13 - 14 | 7819 | 64 | |
NM_001344356.1 | NP_001332643.1 | 13 - 14 | 7816 | 64 | ||
NM_001344357.1 | NP_001332642.1 | 13 - 14 | 7816 | 64 | ||
NM_123447.3 | NP_198898.2 | 13 - 14 | 7816 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |