Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123460.5 | NP_198911.1 | 1 - 2 | 390 | 121 | |
GenBank mRNA | AY046039.1 | - | 1 - 2 | 364 | 121 | |
AY117267.1 | - | 1 - 2 | 306 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |