Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123564.3 | NP_568599.1 | 3 - 4 | 795 | 99 | |
GenBank mRNA | AF424629.1 | - | 3 - 4 | 795 | 99 | |
AY113042.1 | - | 3 - 4 | 351 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |