Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABIL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344432.1 | NP_001330353.1 | 7 - 8 | 858 | 93 | |
NM_001344433.1 | NP_001330352.1 | 6 - 7 | 600 | 93 | ||
NM_123568.5 | NP_199018.1 | 8 - 9 | 900 | 93 | ||
GenBank mRNA | AK175528.1 | - | 8 - 9 | 732 | 93 | |
AK175563.1 | - | 8 - 9 | 701 | 93 | ||
AK176699.1 | - | 8 - 9 | 788 | 93 | ||
AK176912.1 | - | 8 - 9 | 696 | 93 | ||
AY817015.1 | - | 8 - 9 | 692 | 93 | ||
BT024503.1 | - | 8 - 9 | 692 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |