Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123589.4 | NP_199039.1 | 3 - 4 | 1041 | 66 | |
GenBank mRNA | AF361604.1 | - | 3 - 4 | 434 | 66 | |
AY143899.1 | - | 2 - 3 | 363 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |