Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344443.1 | NP_001331624.1 | 13 - 14 | 2462 | 69 | |
NM_123604.3 | NP_199054.1 | 13 - 14 | 2465 | 69 | ||
GenBank mRNA | BT000785.1 | - | 13 - 14 | 2379 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |