Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344444.1 | NP_001318731.1 | 8 - 9 | 2749 | 75 | |
NM_001344445.1 | NP_001330664.1 | 8 - 9 | 2751 | 75 | ||
NM_001344446.1 | NP_001330665.1 | 8 - 9 | 2740 | 75 | ||
NM_001344447.1 | NP_001330668.1 | 8 - 9 | 2748 | 75 | ||
NM_001344448.1 | NP_001330669.1 | 8 - 9 | 2748 | 75 | ||
NM_001344449.1 | NP_001330666.1 | 8 - 9 | 2749 | 75 | ||
NM_001344450.1 | NP_001330667.1 | 7 - 8 | 2571 | 75 | ||
NM_001344451.1 | NP_001330670.1 | 8 - 9 | 2747 | 75 | ||
GenBank mRNA | EU014691.1 | - | 7 - 8 | 2540 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |