Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123624.4 | NP_199074.1 | 6 - 7 | 1045 | 91 | |
GenBank mRNA | BT020312.1 | - | 6 - 7 | 981 | 91 | |
BT020456.1 | - | 6 - 7 | 932 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |