Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123624.4 | NP_199074.1 | 9 - 10 | 1593 | 61 | |
GenBank mRNA | BT020312.1 | - | 9 - 10 | 1529 | 61 | |
BT020456.1 | - | 9 - 10 | 1480 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |