Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123646.4 | NP_568613.1 | 2 - 3 | 682 | 101 | |
GenBank mRNA | AY059898.1 | - | 2 - 3 | 557 | 101 | |
AY093362.1 | - | 2 - 3 | 311 | 101 | ||
DQ122931.1 | - | 2 - 3 | 311 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |