Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123646.4 | NP_568613.1 | 6 - 7 | 1575 | 138 | |
GenBank mRNA | AY059898.1 | - | 6 - 7 | 1450 | 138 | |
AY093362.1 | - | 6 - 7 | 1204 | 138 | ||
DQ122931.1 | - | 6 - 7 | 1204 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |