Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344480.1 | NP_001330706.1 | 1 - 2 | 729 | 85 | |
NM_001344481.1 | NP_001330707.1 | 1 - 2 | 679 | 85 | ||
NM_123698.4 | NP_199146.1 | 1 - 2 | 729 | 85 | ||
GenBank mRNA | AF360257.1 | - | 1 - 2 | 479 | 85 | |
AY040071.1 | - | 1 - 2 | 70 | 85 | ||
AY943854.1 | - | 1 - 2 | 70 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |