Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123726.4 | NP_199173.2 | 2 - 3 | 499 | 81 | |
GenBank mRNA | AK230338.1 | - | 1465 | 81 | ||
AY074343.1 | - | 2 - 3 | 463 | 81 | ||
AY091387.1 | - | 2 - 3 | 288 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |