Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123753.3 | NP_199199.1 | 1 - 2 | 329 | 62 | |
GenBank mRNA | AF134302.1 | - | 1 - 2 | 305 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |