Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YUC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123756.2 | NP_199202.1 | 1 | 1201 | 76 | |
GenBank mRNA | BT029743.1 | - | 1 | 957 | 76 | |
DQ159070.1 | - | 1 | 957 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |