Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203536.1 | NP_001190465.1 | 12 - 13 | 1451 | 133 | |
NM_001203537.1 | NP_001190466.1 | 12 - 13 | 1442 | 133 | ||
NM_123757.5 | NP_199203.1 | 10 - 11 | 1543 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |