Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344547.1 | NP_001331409.1 | 3 - 4 | 647 | 128 | |
NM_001344548.1 | NP_001331410.1 | 3 - 4 | 566 | 128 | ||
NM_001344549.1 | NP_001331408.1 | 2 - 3 | 190 | 128 | ||
NM_123815.1 | NP_199261.1 | 1 - 2 | 262 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |