Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344547.1 | NP_001331409.1 | 10 - 11 | 1471 | 80 | |
NM_001344548.1 | NP_001331410.1 | 10 - 11 | 1390 | 80 | ||
NM_001344549.1 | NP_001331408.1 | 9 - 10 | 1014 | 80 | ||
NM_123815.1 | NP_199261.1 | 8 - 9 | 1086 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |