Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123875.3 | NP_199320.1 | 1 - 2 | 642 | 103 | |
GenBank mRNA | BT010864.1 | - | 1 - 2 | 503 | 103 | |
BT012618.1 | - | 1 - 2 | 481 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |