Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123907.4 | NP_568644.1 | 1 - 2 | 494 | 120 | |
GenBank mRNA | AY042832.1 | - | 1 - 2 | 442 | 120 | |
BT006321.1 | - | 1 - 2 | 397 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |