Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123958.3 | NP_568652.1 | 9 - 10 | 1884 | 75 | |
GenBank mRNA | AB073173.1 | - | 9 - 10 | 1837 | 75 | |
AF492761.1 | - | 9 - 10 | 1755 | 75 | ||
AY034945.1 | - | 9 - 10 | 1815 | 75 | ||
AY150456.1 | - | 9 - 10 | 1755 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |