Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125908.1 | NP_001119380.1 | 2 - 3 | 444 | 102 | |
NM_001344651.1 | NP_001331230.1 | 2 - 3 | 429 | 102 | ||
NM_123962.3 | NP_199406.1 | 2 - 3 | 429 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |