Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344656.1 | NP_001331060.1 | 2 - 3 | 1183 | 96 | |
NM_123973.4 | NP_199417.1 | 4 - 5 | 1413 | 96 | ||
GenBank mRNA | AY150511.1 | - | 4 - 5 | 1293 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |