Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123990.3 | NP_568658.1 | 8 - 9 | 1697 | 81 | |
GenBank mRNA | AY084267.1 | - | 8 - 9 | 1556 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |