Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123993.3 | NP_199436.1 | 8 - 9 | 2326 | 130 | |
GenBank mRNA | AK176505.1 | - | 8 - 9 | 1526 | 130 | |
AK220637.1 | - | 8 - 9 | 1526 | 130 | ||
BT046198.1 | - | 8 - 9 | 1481 | 130 | ||
M86990.1 | - | 8 - 9 | 1500 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |