Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124024.2 | NP_199466.1 | 5 - 6 | 1041 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |