Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124150.5 | NP_199587.1 | 8 - 9 | 750 | 61 | |
GenBank mRNA | AK118640.1 | - | 8 - 9 | 617 | 61 | |
BT005294.1 | - | 8 - 9 | 602 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |