Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124152.3 | NP_568688.1 | 6 - 7 | 752 | 70 | |
GenBank mRNA | AY056202.1 | - | 6 - 7 | 700 | 70 | |
AY091394.1 | - | 6 - 7 | 635 | 70 | ||
KJ138647.1 | - | 6 - 7 | 635 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |