Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBOHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124165.3 | NP_199602.1 | 3 - 4 | 1376 | 104 | |
GenBank mRNA | AF424625.1 | - | 3 - 4 | 974 | 104 | |
BT002651.1 | - | 3 - 4 | 832 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |