Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBOHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124165.3 | NP_199602.1 | 7 - 8 | 3193 | 71 | |
GenBank mRNA | AF424625.1 | - | 7 - 8 | 2791 | 71 | |
BT002651.1 | - | 7 - 8 | 2649 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |