Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124184.2 | NP_199621.2 | 4 - 5 | 761 | 84 | |
GenBank mRNA | BT002919.1 | - | 4 - 5 | 632 | 84 | |
BT005152.1 | - | 4 - 5 | 599 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |