Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124205.4 | NP_199641.1 | 3 - 4 | 1105 | 65 | |
GenBank mRNA | AY049265.1 | - | 3 - 4 | 1081 | 65 | |
AY065428.1 | - | 3 - 4 | 1047 | 65 | ||
AY090283.1 | - | 3 - 4 | 1017 | 65 | ||
AY096379.1 | - | 3 - 4 | 1017 | 65 | ||
U72351.1 | - | 3 - 4 | 1052 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |