Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203563.2 | NP_001190492.1 | 9 - 10 | 1264 | 153 | |
NM_124236.3 | NP_199671.1 | 9 - 10 | 1115 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |