Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161298.2 | NP_001154770.1 | 8 - 9 | 771 | 107 | |
NM_001344803.1 | NP_001330159.1 | 7 - 8 | 712 | 107 | ||
NM_001344804.1 | NP_001330160.1 | 7 - 8 | 659 | 107 | ||
NM_124249.3 | NP_199683.2 | 7 - 8 | 944 | 107 | ||
GenBank mRNA | BT012214.1 | - | 7 - 8 | 608 | 107 | |
BT020567.1 | - | 5 - 6 | 443 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |