Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036965.2 | NP_001032042.1 | 13 - 14 | 2012 | 112 | |
NM_001203568.1 | NP_001190497.1 | 14 - 15 | 2445 | 112 | ||
NM_124280.4 | NP_199714.2 | 13 - 14 | 2124 | 112 | ||
GenBank mRNA | AY080830.1 | - | 13 - 14 | 1941 | 112 | |
AY142605.1 | - | 13 - 14 | 1841 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |