Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036965.2 | NP_001032042.1 | 2 - 3 | 446 | 72 | |
NM_001203568.1 | NP_001190497.1 | 3 - 4 | 879 | 72 | ||
NM_124280.4 | NP_199714.2 | 2 - 3 | 558 | 72 | ||
GenBank mRNA | AY080830.1 | - | 2 - 3 | 375 | 72 | |
AY142605.1 | - | 2 - 3 | 275 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |