Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344824.1 | NP_001331823.1 | 13 - 14 | 2408 | 102 | |
NM_001344825.1 | NP_001331822.1 | 13 - 14 | 2320 | 102 | ||
NM_124296.3 | NP_199730.1 | 14 - 15 | 2785 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |