Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124301.1 | NP_199735.1 | 1 - 2 | 192 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |