Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124308.2 | NP_199742.1 | 7 - 8 | 1109 | 84 | |
GenBank mRNA | DQ447054.1 | - | 7 - 8 | 953 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |