Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124329.3 | NP_199763.1 | 1 - 2 | 821 | 98 | |
GenBank mRNA | AF442397.1 | - | 1 - 2 | 579 | 98 | |
AY063870.1 | - | 1 - 2 | 693 | 98 | ||
AY091224.1 | - | 1 - 2 | 579 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |