Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124335.3 | NP_199768.1 | 11 - 12 | 1075 | 103 | |
GenBank mRNA | AK228156.1 | - | 11 - 12 | 995 | 103 | |
AM745095.1 | - | 11 - 12 | 931 | 103 | ||
AY140065.1 | - | 11 - 12 | 991 | 103 | ||
BT003398.1 | - | 11 - 12 | 931 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |